missing translation for 'onlineSavingsMsg'
Learn More

Morpholine, TCI America™

Catalog No. M046525ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M046525ML 25 mL
M0465500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M046525ML Supplier TCI America Supplier No. M046525ML

Chemical Identifiers

CAS 110-91-8
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.12
MDL Number MFCD00005972
InChI Key YNAVUWVOSKDBBP-UHFFFAOYSA-N
Synonym 1-oxa-4-azacyclohexane, tetrahydro-1,4-oxazine, diethylene oximide, diethylenimide oxide, diethyleneimide oxide, drewamine, diethylene imidoxide, tetrahydro-p-oxazine, tetrahydro-2h-1,4-oxazine, p-isoxazine, tetrahydro
PubChem CID 8083
ChEBI CHEBI:34856
IUPAC Name morpholine
SMILES C1COCCN1

Specifications

Melting Point -5°C
Color Yellow
pH 11 (25% H2O soln.)
Boiling Point 129°C
Quantity 25 mL
UN Number 2054
Formula Weight 87.12
Percent Purity ≥99.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Morpholine
EINECSNumber (5)-0859
RTECSNumber QD6475000
TSCA Yes