missing translation for 'onlineSavingsMsg'
Learn More

Morpholine, ACS Reagent Grade, Ricca Chemical

Catalog No. RSOM0200500
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RSOM0200500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RSOM0200500 Supplier Ricca Chemical Company Supplier No. RSOM0200500C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 110-91-8
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.12
MDL Number MFCD00005972
InChI Key YNAVUWVOSKDBBP-UHFFFAOYSA-N
Synonym 1-oxa-4-azacyclohexane, tetrahydro-1,4-oxazine, diethylene oximide, diethylenimide oxide, diethyleneimide oxide, drewamine, diethylene imidoxide, tetrahydro-p-oxazine, tetrahydro-2h-1,4-oxazine, p-isoxazine, tetrahydro
PubChem CID 8083
ChEBI CHEBI:34856
IUPAC Name morpholine
SMILES C1COCCN1

Specifications

CAS Min % 98.0
CAS Max % 100.0
Boiling Point Approximately 126°C to 130°C
Packaging Amber Glass Bottle
Quantity 500 mL
Percent Purity Min. 99%
Grade ACS
Physical Form Liquid
Chemical Name or Material Morpholine
ShelfLife 60 months