missing translation for 'onlineSavingsMsg'
Learn More

Morpholine, 99+%, extra pure

Catalog No. AC158680010
Change view
Click to view available options
Quantity:
5 mL
1 L
2.5 L
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC158680010 1 L Glass Bottle
AC158680050 5 mL Glass Bottle
AC158680025 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC158680010 Supplier Thermo Scientific Chemicals Supplier No. 158680010
Only null left

Morpholine, 99+%, C4H9NO, CAS Number-110-91-8, 1-oxa-4-azacyclohexane, diethyleneimide oxide, diethylene imidoxide, diethylene oximide, diethylenimide oxide, drewamine, p-isoxazine, tetrahydro, tetrahydro-1,4-oxazine, tetrahydro-2h-1,4-oxazine, tetrahydro-p-oxazine, 1L, 99% min. (GC), 27, 5 | CAS: 110-91-8 | C4H9NO | 87.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 110-91-8
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.12
MDL Number MFCD00005972
InChI Key YNAVUWVOSKDBBP-UHFFFAOYSA-N
Synonym 1-oxa-4-azacyclohexane, tetrahydro-1,4-oxazine, diethylene oximide, diethylenimide oxide, diethyleneimide oxide, drewamine, diethylene imidoxide, tetrahydro-p-oxazine, tetrahydro-2h-1,4-oxazine, p-isoxazine, tetrahydro
PubChem CID 8083
ChEBI CHEBI:34856
IUPAC Name morpholine
SMILES C1COCCN1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -5.0°C
Density 0.9900g/mL
Boiling Point 126.0°C to 130.0°C
Flash Point 32°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass Bottle
Refractive Index 1.4530 to 1.4550
Quantity 1 L
Beilstein 27, 5
Fieser 01,705; 11,352; 13,14
Specific Gravity 0.99
Merck Index 15, 6362
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol, ethanol, acetone, benzene,, ether, castor oil, ethylene glycol, 2-hexanone,, linseed oil, turpentine, pine oil
Viscosity 2.23 mPa.s (20°C)
Formula Weight 87.12
Percent Purity 99+%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material Morpholine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if inhaled.
Harmful if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN (
EINECSNumber 203-815-1

RUO – Research Use Only