Learn More
Morpholine-4-carboxamidine hydrobromide, 98%
CAS: 157415-17-3 | C5H12BrN3O | 210.075 g/mol
Supplier: Thermo Scientific Chemicals A1998514
Description
It is used as an active pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as an active pharmaceutical intermediate.
Solubility
Soluble in water.
Notes
Hygroscopic. Store away from oxidizing agents, water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
| Morpholine-4-carboxamidine hydrobromide | |
| ∼155°C to 165°C | |
| MFCD00051994 | |
| 7389038 | |
| morpholine-4-carboximidamide hydrobromide, morpholinoformamidine hydrobromide, 4-morpholinylformamidine hydrobromide, morpholine-4-carboxamidine hydrobromide, 4-morpholinecarboximidamide, monohydrobromide, acmc-20adpt, morpholine-4-carboxamidine, bromide, morpholin-4-carboxamidine hydrobromide | |
| IOHSDHMCZXCESZ-UHFFFAOYSA-N | |
| morpholine-4-carboximidamide;hydrobromide | |
| 2806564 | |
| 98% |
| 157415-17-3 | |
| C5H12BrN3O | |
| 25 g | |
| Hygroscopic | |
| Soluble in water. | |
| C1COCCN1C(=N)N.Br | |
| 210.075 | |
| 210.08 |
Chemical Identifiers
| 157415-17-3 | |
| 210.075 | |
| IOHSDHMCZXCESZ-UHFFFAOYSA-N | |
| 2806564 | |
| C1COCCN1C(=N)N.Br |
| C5H12BrN3O | |
| MFCD00051994 | |
| morpholine-4-carboximidamide hydrobromide, morpholinoformamidine hydrobromide, 4-morpholinylformamidine hydrobromide, morpholine-4-carboxamidine hydrobromide, 4-morpholinecarboximidamide, monohydrobromide, acmc-20adpt, morpholine-4-carboxamidine, bromide, morpholin-4-carboxamidine hydrobromide | |
| morpholine-4-carboximidamide;hydrobromide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only