missing translation for 'onlineSavingsMsg'
Learn More

Morpholine-4-carboxamidine hydrobromide, 98%

Catalog No. AAA1998506
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1998506 5 g
AAA1998514 25 g
AAA1998522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1998506 Supplier Thermo Scientific Chemicals Supplier No. A1998506

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 157415-17-3 | C5H12BrN3O | 210.075 g/mol

It is used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Hygroscopic. Store away from oxidizing agents, water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 157415-17-3
Molecular Formula C5H12BrN3O
Molecular Weight (g/mol) 210.075
MDL Number MFCD00051994
InChI Key IOHSDHMCZXCESZ-UHFFFAOYSA-N
Synonym morpholine-4-carboximidamide hydrobromide, morpholinoformamidine hydrobromide, 4-morpholinylformamidine hydrobromide, morpholine-4-carboxamidine hydrobromide, 4-morpholinecarboximidamide, monohydrobromide, acmc-20adpt, morpholine-4-carboxamidine, bromide, morpholin-4-carboxamidine hydrobromide
PubChem CID 2806564
IUPAC Name morpholine-4-carboximidamide;hydrobromide
SMILES C1COCCN1C(=N)N.Br

Specifications

Melting Point ∼155°C to 165°C
Quantity 5 g
Beilstein 7389038
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 210.08
Percent Purity 98%
Chemical Name or Material Morpholine-4-carboxamidine hydrobromide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only