missing translation for 'onlineSavingsMsg'
Learn More

Morpholin-3-one, 98%

Catalog No. AC460530250
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC460530250 25 g
AC460530010 1 g
AC460530050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC460530250 Supplier Thermo Scientific Chemicals Supplier No. 460530250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 109-11-5 | C4H7NO2 | 101.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 109-11-5
Molecular Formula C4H7NO2
Molecular Weight (g/mol) 101.11
MDL Number MFCD00631009
InChI Key VSEAAEQOQBMPQF-UHFFFAOYSA-N
Synonym 3-morpholinone, 3-ketomorpholine, morpholinone, morpholine-3-one, 3-oxomorpholine, 3-oxo-morpholine, acmc-1bpa5, ksc180o3n, morpholin-3-one; 3-morpholone
PubChem CID 66953
IUPAC Name morpholin-3-one
SMILES O=C1COCCN1

Specifications

Melting Point 106.0°C to 108.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 25 g
Formula Weight 101.11
Percent Purity 98%
Chemical Name or Material Morpholin-3-one
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. Causes skin irritation. May cause an allergic skin reaction. Causes serious eye damage. May cause respiratory irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF INHALED. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
EINECSNumber 203-647-9

RUO – Research Use Only