missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Morin, Saturated Methanolic, Ricca Chemical
Supplier: Ricca Chemical Company R5217000120C
Specifications
| Alcohol | |
| CH4O | |
| 120 mL | |
| Solubility in Water: Infinite | |
| CO | |
| 32.04 |
| Amber Glass Bottle | |
| MFCD00004595 | |
| methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
| OKKJLVBELUTLKV-UHFFFAOYSA-N | |
| methanol |
Chemical Identifiers
| 0.0 |
Safety and Handling
Recommended Storage : Flammable