missing translation for 'onlineSavingsMsg'
Learn More

Morin Hydrate 90.0+%, TCI America™

Catalog No. P00411G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P00411G 1 g
P00415G 5 g
P004125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P00411G Supplier TCI America Supplier No. P00411G

Chemical Identifiers

CAS 654055-01-3
Molecular Formula C15H10O7
Molecular Weight (g/mol) 302.24
MDL Number MFCD00217054
InChI Key YXOLAZRVSSWPPT-UHFFFAOYSA-N
Synonym morin hydrate, morinhydrate, bois d,arc hydrate, 2-2,4-dihydroxyphenyl-3,5,7-trihydroxy-4h-chromen-4-one hydrate, morin hydrate, powder, morin flavonol, morin hydrate aurantica, 2-2,4-dihydroxyphenyl-3,5,7-trihydroxy-chromen-4-one hydrate, 2 inverted exclamation marka,3,4 inverted exclamation marka,5,7-pentahydroxyflavone
PubChem CID 16219651
IUPAC Name 2-(2,4-dihydroxyphenyl)-3,5,7-trihydroxy-4H-chromen-4-one
SMILES OC1=CC(O)=C(C=C1)C1=C(O)C(=O)C2=C(O)C=C(O)C=C2O1

Specifications

Quantity 1 g
UN Number 3077
Formula Weight 302.24
Percent Purity ≥90.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Morin Hydrate
EINECSNumber (5)-5230
RTECSNumber LK8749000
TSCA Yes