missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MOPS sodium salt, 97%
CAS: 71119-22-7 | C7H14NNaO4S | 231.242 g/mol
$80.15 - $195.95
Chemical Identifiers
| CAS | 71119-22-7 |
|---|---|
| Molecular Formula | C7H14NNaO4S |
| MDL Number | MFCD00064350 |
| InChI Key | MWEMXEWFLIDTSJ-UHFFFAOYSA-M |
| Synonym | mops sodium salt, sodium 3-morpholinopropanesulfonate, 4-morpholinepropanesulfonic acid, sodium salt, sodium 3-morpholin-4-ylpropane-1-sulfonate, 3-n-morpholino propanesulfonic acid sodium salt, 3-4-morpholino propanesulfonic acid sodium salt, 4-morpholinepropanesulfonic acid, sodium salt 1:1, 4-morpholinepropanesulfonic acid sodium salt, sodium 3-morpholin-4-yl propane-1-sulfonate, 3-morpholin-4-ylpropanesulfonic acid, sodium salt |
| PubChem CID | 3859613 |
| IUPAC Name | sodium;3-morpholin-4-ylpropane-1-sulfonate |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1721414
|
Thermo Scientific Chemicals
A1721414 |
25 g |
Each for $80.15
|
|
|||||
|
AAA1721422
|
Thermo Scientific Chemicals
A1721422 |
100 g |
Each for $195.95
|
|
|||||
Description
3-(N-Morpholino)propanesulfonic Acid, Na; an analog to MES used in polyacrylamide gel electrophoresis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-(N-Morpholino)propanesulfonic Acid, Na; an analog to MES used in polyacrylamide gel electrophoresis.
Solubility
Soluble in water (523 g/L at 20°C).
Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
Chemical Identifiers
| 71119-22-7 | |
| MFCD00064350 | |
| mops sodium salt, sodium 3-morpholinopropanesulfonate, 4-morpholinepropanesulfonic acid, sodium salt, sodium 3-morpholin-4-ylpropane-1-sulfonate, 3-n-morpholino propanesulfonic acid sodium salt, 3-4-morpholino propanesulfonic acid sodium salt, 4-morpholinepropanesulfonic acid, sodium salt 1:1, 4-morpholinepropanesulfonic acid sodium salt, sodium 3-morpholin-4-yl propane-1-sulfonate, 3-morpholin-4-ylpropanesulfonic acid, sodium salt | |
| sodium;3-morpholin-4-ylpropane-1-sulfonate |
| C7H14NNaO4S | |
| MWEMXEWFLIDTSJ-UHFFFAOYSA-M | |
| 3859613 |
Specifications
| ∽260°C (decomposition) | |
| 25 g | |
| C7H14NNaO4S | |
| mops sodium salt, sodium 3-morpholinopropanesulfonate, 4-morpholinepropanesulfonic acid, sodium salt, sodium 3-morpholin-4-ylpropane-1-sulfonate, 3-n-morpholino propanesulfonic acid sodium salt, 3-4-morpholino propanesulfonic acid sodium salt, 4-morpholinepropanesulfonic acid, sodium salt 1:1, 4-morpholinepropanesulfonic acid sodium salt, sodium 3-morpholin-4-yl propane-1-sulfonate, 3-morpholin-4-ylpropanesulfonic acid, sodium salt | |
| MWEMXEWFLIDTSJ-UHFFFAOYSA-M | |
| sodium;3-morpholin-4-ylpropane-1-sulfonate | |
| 3859613 | |
| MOPS sodium salt |
| 71119-22-7 | |
| 97% | |
| MFCD00064350 | |
| Soluble in water (523g/L at 20°C). | |
| C1COCCN1CCCS(=O)(=O)[O-].[Na+] | |
| 231.242 | |
| 231.25 |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only