missing translation for 'onlineSavingsMsg'
Learn More

MOPS sodium salt, 97%

Catalog No. AAA1721422
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1721422 100 g
AAA1721414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1721422 Supplier Thermo Scientific Chemicals Supplier No. A1721422
Only null left

CAS: 71119-22-7 | C7H14NNaO4S | 231.242 g/mol

3-(N-Morpholino)propanesulfonic Acid, Na; an analog to MES used in polyacrylamide gel electrophoresis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(N-Morpholino)propanesulfonic Acid, Na; an analog to MES used in polyacrylamide gel electrophoresis.

Solubility
Soluble in water (523 g/L at 20°C).

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.

Chemical Identifiers

CAS 71119-22-7
Molecular Formula C7H14NNaO4S
MDL Number MFCD00064350
InChI Key MWEMXEWFLIDTSJ-UHFFFAOYSA-M
Synonym mops sodium salt, sodium 3-morpholinopropanesulfonate, 4-morpholinepropanesulfonic acid, sodium salt, sodium 3-morpholin-4-ylpropane-1-sulfonate, 3-n-morpholino propanesulfonic acid sodium salt, 3-4-morpholino propanesulfonic acid sodium salt, 4-morpholinepropanesulfonic acid, sodium salt 1:1, 4-morpholinepropanesulfonic acid sodium salt, sodium 3-morpholin-4-yl propane-1-sulfonate, 3-morpholin-4-ylpropanesulfonic acid, sodium salt
PubChem CID 3859613
IUPAC Name sodium;3-morpholin-4-ylpropane-1-sulfonate

Specifications

Melting Point ∽260°C (decomposition)
Quantity 100 g
Assay Percent Range 97%
Solubility Information Soluble in water (523g/L at 20°C).
SMILES C1COCCN1CCCS(=O)(=O)[O-].[Na+]
Molecular Weight (g/mol) 231.242
Formula Weight 231.25
Chemical Name or Material MOPS sodium salt
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only