missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma MOPS SDS Running Buffer Powder

Catalog No. 11580586
Change view
Click to view available options
Physical Form:
Powder
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Physical Form
11580586 Powder
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11580586 Supplier MilliporeSigma Supplier No. MPM0PS

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MOPS SDS running buffer powder for mPAGE™ Bis-Tris gels

  • MOPS SDS running buffer powder for mPAGE Bis-Tris gels.
  • 5 packets. Each packet makes 1 liter of running buffer.
  • Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Specifications

Chemical Name or Material MOPS SDS running buffer powder for mPAGE™ Bis-Tris gels
Physical Form Powder
Quantity 5 Pk.