missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ MOPS, Free Acid, Molecular Biology Grade, Calbiochem™,
Specifications
Specifications
| Quantity | 100 g |
| DNase | None Detected |
| Assay Percent Range | ≥99.0% |
| Protease | None Detected |
| Solubility Information | Soluble in cold water |
| Formula Weight | 209.3 |
| Grade | Potentiometry |
| Chemical Name or Material | MOPS, Free Acid, Molecular Biology Grade |
Safety and Handling
| Recommended Storage | 15°C to 30°C (Do not freeze). |