missing translation for 'onlineSavingsMsg'
Learn More

MOPS, Biological Buffer, 99%, Spectrum™ Chemical

Catalog No. p-7157880
Change view
Click to view available options
Quantity:
100 g
5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604695 100 g Poly Bottle
18604697 5 kg Poly Pail
18604699 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18604695 Supplier Spectrum Chemical Mfg Cor Supplier No. MO102100GM
Only null left

MO102, 1132-61-2, C7H15NO4S

Spectrum™ Chemical MOPS, Biological Buffer

Specifications

CAS 1132-61-2
CAS Max % 1
Loss on Drying 0.01
Packaging Poly Bottle
Quantity 100 g
Percent Purity 99%
Grade Reagent