missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monosodium Succinate 98.0+%, TCI America™
$74.56 - $195.73
Chemical Identifiers
| CAS | 2922-54-5 |
|---|---|
| Molecular Formula | C4H5NaO4 |
| Molecular Weight (g/mol) | 140.07 |
| MDL Number | MFCD00050993 |
| InChI Key | KZQSXALQTHVPDQ-UHFFFAOYSA-M |
| Synonym | Sodium Bisuccinate, Sodium Hydrogen Succinate, Succinic Acid Monosodium Salt |
| PubChem CID | 23690498 |
| IUPAC Name | sodium;4-hydroxy-4-oxobutanoate |
| SMILES | C(CC(=O)[O-])C(=O)O.[Na+] |
Chemical Identifiers
| 2922-54-5 | |
| 140.07 | |
| KZQSXALQTHVPDQ-UHFFFAOYSA-M | |
| 23690498 | |
| C(CC(=O)[O-])C(=O)O.[Na+] |
| C4H5NaO4 | |
| MFCD00050993 | |
| Sodium Bisuccinate, Sodium Hydrogen Succinate, Succinic Acid Monosodium Salt | |
| sodium;4-hydroxy-4-oxobutanoate |
Specifications
| 2922-54-5 | |
| C4H5NaO4 | |
| 25 g | |
| KZQSXALQTHVPDQ-UHFFFAOYSA-M | |
| sodium;4-hydroxy-4-oxobutanoate | |
| 23690498 | |
| ≥98.0% (T) | |
| Monosodium Succinate |
| White | |
| MFCD00050993 | |
| Sodium Bisuccinate, Sodium Hydrogen Succinate, Succinic Acid Monosodium Salt | |
| C(CC(=O)[O-])C(=O)O.[Na+] | |
| 140.07 | |
| 140.07 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-0911
RTECSNumber : WN0500000
TSCA : No