missing translation for 'onlineSavingsMsg'
Learn More

Monosodium Succinate 98.0+%, TCI America™

Catalog No. S0106500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0106500G 500 g
S010625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S0106500G Supplier TCI America Supplier No. S0106500G

Chemical Identifiers

CAS 2922-54-5
Molecular Formula C4H5NaO4
Molecular Weight (g/mol) 140.07
MDL Number MFCD00050993
InChI Key KZQSXALQTHVPDQ-UHFFFAOYSA-M
Synonym Sodium Bisuccinate, Sodium Hydrogen Succinate, Succinic Acid Monosodium Salt
PubChem CID 23690498
IUPAC Name sodium;4-hydroxy-4-oxobutanoate
SMILES C(CC(=O)[O-])C(=O)O.[Na+]

Specifications

Color White
Quantity 500 g
Formula Weight 140.07
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Monosodium Succinate
EINECSNumber (2)-0911
RTECSNumber WN0500000
TSCA No