missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monosodium Succinate 98.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 25 g |
| Formula Weight | 140.07 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Monosodium Succinate |
Safety and Handling
| EINECSNumber | (2)-0911 |
| RTECSNumber | WN0500000 |
| TSCA | No |