missing translation for 'onlineSavingsMsg'
Learn More

Monosodium Bromoisocyanurate Hydrate 95.0+%, TCI America™

Catalog No. B214825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B214825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B214825G Supplier TCI America Supplier No. B214825G
Only null left

Chemical Identifiers

CAS 164918-61-0
Molecular Formula C3H2BrN3O3
MDL Number MFCD04037229
Synonym Bromoisocyanuric Acid Monosodium Salt

Specifications

Color White
Quantity 25 g
Formula Weight 229.95
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Monosodium Bromoisocyanurate Hydrate