missing translation for 'onlineSavingsMsg'
Learn More

Monomethyl Suberate 98.0+%, TCI America™

$67.38 - $165.56

Chemical Identifiers

CAS 3946-32-5
Molecular Formula C9H15O4
Molecular Weight (g/mol) 187.22
MDL Number MFCD00004427
InChI Key KOVPXZDUVJGGFU-UHFFFAOYSA-M
Synonym suberic acid monomethyl ester, monomethyl suberate, methyl hydrogen suberate, subericacidmonomethylester, octanedioic acid monomethyl ester, mono-methyl suberate, acmc-1ckw1, ksc492m6f, 7-methoxycarbonylheptanoic acid, octanedioic acid 1-methyl ester
PubChem CID 554191
IUPAC Name 8-methoxy-8-oxooctanoate
SMILES COC(=O)CCCCCCC([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
S04405G
View Documents
TCI America
S04405G
5 g
Each for $67.38
Only null left
 
S044025G
View Documents
TCI America
S044025G
25 g
Each for $165.56
Only null left
 
Description

Description

Specifications

Chemical Identifiers

3946-32-5
187.22
KOVPXZDUVJGGFU-UHFFFAOYSA-M
554191
COC(=O)CCCCCCC([O-])=O
C9H15O4
MFCD00004427
suberic acid monomethyl ester, monomethyl suberate, methyl hydrogen suberate, subericacidmonomethylester, octanedioic acid monomethyl ester, mono-methyl suberate, acmc-1ckw1, ksc492m6f, 7-methoxycarbonylheptanoic acid, octanedioic acid 1-methyl ester
8-methoxy-8-oxooctanoate

Specifications

3946-32-5
135°C
MFCD00004427
suberic acid monomethyl ester, monomethyl suberate, methyl hydrogen suberate, subericacidmonomethylester, octanedioic acid monomethyl ester, mono-methyl suberate, acmc-1ckw1, ksc492m6f, 7-methoxycarbonylheptanoic acid, octanedioic acid 1-methyl ester
COC(=O)CCCCCCC([O-])=O
187.22
188.22
Crystalline Lumps
White-Yellow
C9H15O4
5 g
KOVPXZDUVJGGFU-UHFFFAOYSA-M
8-methoxy-8-oxooctanoate
554191
≥98.0% (GC,T)
Monomethyl Suberate
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents