missing translation for 'onlineSavingsMsg'
Learn More

Monomethyl Suberate 98.0+%, TCI America™

Catalog No. S04405G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S04405G 5 g
S044025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S04405G Supplier TCI America Supplier No. S04405G

Chemical Identifiers

CAS 3946-32-5
Molecular Formula C9H15O4
Molecular Weight (g/mol) 187.22
MDL Number MFCD00004427
InChI Key KOVPXZDUVJGGFU-UHFFFAOYSA-M
Synonym suberic acid monomethyl ester, monomethyl suberate, methyl hydrogen suberate, subericacidmonomethylester, octanedioic acid monomethyl ester, mono-methyl suberate, acmc-1ckw1, ksc492m6f, 7-methoxycarbonylheptanoic acid, octanedioic acid 1-methyl ester
PubChem CID 554191
IUPAC Name 8-methoxy-8-oxooctanoate
SMILES COC(=O)CCCCCCC([O-])=O

Specifications

Color White-Yellow
Boiling Point 135°C
Quantity 5 g
Formula Weight 188.22
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material Monomethyl Suberate