missing translation for 'onlineSavingsMsg'
Learn More

Monomethyl Sebacate 98.0+%, TCI America™

Catalog No. p-7135877
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S03955G 5 g
S039525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S03955G Supplier TCI America Supplier No. S03955G

Chemical Identifiers

CAS 818-88-2
Molecular Formula C11H20O4
Molecular Weight (g/mol) 216.28
MDL Number MFCD00004439
InChI Key OSYQOBUUFRGFNG-UHFFFAOYSA-N
Synonym Decanedioic Acid Monomethyl Ester, Methyl Hydrogen Sebacate, Sebacic Acid Monomethyl Ester
PubChem CID 69957
IUPAC Name 10-methoxy-10-oxodecanoic acid
SMILES COC(=O)CCCCCCCCC(O)=O

Specifications

Melting Point 42°C
Color White
Quantity 5 g
Formula Weight 216.28
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Monomethyl Sebacate