missing translation for 'onlineSavingsMsg'
Learn More

Monomethyl monopotassium malonate, 98%, Thermo Scientific™

Catalog No. AC269550250
Change view
Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC269550250 25g Glass bottle
AC269551000 100g Glass bottle
AC269555000 500g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC269550250 Supplier Thermo Scientific Chemicals Supplier No. 269550250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 38330-80-2
Molecular Formula C4H5O4·K
Molecular Weight (g/mol) 156.18
MDL Number MFCD00014021
InChI Key WWTULTKUWBKVGV-UHFFFAOYSA-M
Synonym potassium 3-methoxy-3-oxopropanoate, methyl potassium malonate, potassium methyl malonate, propanedioic acid, monomethyl ester, potassium salt, potassium monomethyl malonate, monomethyl malonate potassium salt, hydrogen methyl malonate potassium salt, monomethylpotassiummalonate, acmc-209vp4, ksc226a9d
PubChem CID 2724687
IUPAC Name potassium;3-methoxy-3-oxopropanoate
SMILES COC(=O)CC(=O)[O-].[K+]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula COOKCH2COOCH3
Quantity 25g
Fieser 15,207
Formula Weight 156.18
Percent Purity 98%
Chemical Name or Material Monomethyl monopotassium malonate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 253-886-8

RUO – Research Use Only