missing translation for 'onlineSavingsMsg'
Learn More

Monomethyl Maleate 90.0+%, TCI America™

Catalog No. M0624500ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0624500ML 500 mL
M062425ML 25 mL
M0624100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M0624500ML Supplier TCI America Supplier No. M0624500ML
Only null left

Chemical Identifiers

CAS 3052-50-4
Molecular Formula C5H6O4
Molecular Weight (g/mol) 130.10
MDL Number MFCD00045905,MFCD00063174
InChI Key NKHAVTQWNUWKEO-IHWYPQMZSA-N
Synonym Maleic Acid Monomethyl Ester, Methyl Hydrogen Maleate
PubChem CID 5354456
IUPAC Name (2Z)-4-methoxy-4-oxobut-2-enoic acid
SMILES COC(=O)\C=C/C(O)=O

Specifications

Color Yellow
Quantity 500 mL
Formula Weight 130.10
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material Monomethyl Maleate
EINECSNumber (2)-1105&(2)-1142
TSCA Yes
Recommended Storage Refrigerator