missing translation for 'onlineSavingsMsg'
Learn More

Monomethyl Fumarate 98.0+%, TCI America™

Catalog No. M24135G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M24135G 5 g
M241325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M24135G Supplier TCI America Supplier No. M24135G

Chemical Identifiers

CAS 2756-87-8
Molecular Formula C5H6O4
Molecular Weight (g/mol) 130.099
MDL Number MFCD00063174
InChI Key NKHAVTQWNUWKEO-NSCUHMNNSA-N
Synonym monomethyl fumarate, 4-methoxy-4-oxobut-2-enoic acid, methyl hydrogen fumarate, mono-methyl fumarate, fumaric acid monomethyl ester, unii-45iub1px8r, monomethylfumarate, 45iub1px8r, e-4-methoxy-4-oxobut-2-enoic acid, 2e-4-methoxy-4-oxobut-2-enoic acid
PubChem CID 5369209
IUPAC Name (E)-4-methoxy-4-oxobut-2-enoic acid
SMILES COC(=O)C=CC(=O)O

Specifications

Melting Point 147°C
Color White
Quantity 5 g
Formula Weight 130.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Monomethyl Fumarate
EINECSNumber (2)-1142
TSCA No