missing translation for 'onlineSavingsMsg'
Learn More

Monomethyl 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylate 98.0+%, TCI America™

Catalog No. M24935G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M24935G 5 g
M249325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M24935G Supplier TCI America Supplier No. M24935G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 74936-72-4
Molecular Formula C16H16N2O6
Molecular Weight (g/mol) 332.312
MDL Number MFCD00475478
InChI Key JPXPPUOCSLMCHK-UHFFFAOYSA-N
Synonym 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic Acid Monomethyl Ester
PubChem CID 173322
IUPAC Name 5-methoxycarbonyl-2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3-carboxylic acid
SMILES CC1=C(C(C(=C(N1)C)C(=O)OC)C2=CC(=CC=C2)[N+](=O)[O-])C(=O)O

Specifications

Melting Point 203°C
Color White-Yellow
Quantity 5 g
Formula Weight 332.31
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Monomethyl 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylate