missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Covidien Monoject™ Nonsilicone Coated Tubes with Gray Stoppers
Description
- Sterilization applies to the tube and stopper interior only, and not to the outside of the tube or to the packaging
- Sterilized by gamma radiation
- Used for laboratory procedures requiring plasma or whole blood and chemistry procedures where glycolytic inhibition of specimen is required
Specifications
Specifications
| Color | Gray |
| Product Type | Non-silicone Coated Tube |
| Additive | Powder Additive: 20 mg Potassium Oxalate, 25 mg Sodium Fluoride |
| Capacity (Metric) | 10 mL |
| Closure Color | Gray |
| Tube Size | 16 x 100 mm |
| For Use With (Application) | Blood collection |