missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monoethylglycinexylidine, ≥98% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 7728-40-7 |
| Linear Formula | C12H18N2 O |
| MDL Number | MFCD00098991 |
| Synonym | MEGX; MGEX; Monoethylglycinexylidide; N-(2,6-dimethylphenyl)-2-(ethylamino)acetamide; Norlidocaine |
| Type | Analytical Standard |
| Quantity | 25 mg |
| Formula Weight | 206.28 |
| Percent Purity | ≥98% (HPLC) |
| Grade | Analytical Standard |