missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monoethyl Potassium Malonate 98.0+%, TCI America™
$94.33 - $377.33
Chemical Identifiers
| CAS | 6148-64-7 |
|---|---|
| Molecular Formula | C5H7KO4 |
| Molecular Weight (g/mol) | 170.205 |
| MDL Number | MFCD00035603 |
| InChI Key | WVUCPRGADMCTBN-UHFFFAOYSA-M |
| Synonym | ethyl potassium malonate, potassium 3-ethoxy-3-oxopropanoate, potassium ethyl malonate, potassium monoethyl malonate, monoethyl malonate potassium salt, malonic acid monoethyl ester potassium salt, monoethyl potassium malonate, ethyl malonate potassium salt, propanedioic acid, monoethyl ester, potassium salt, ethylpotassiummalonate |
| PubChem CID | 3446434 |
| IUPAC Name | potassium;3-ethoxy-3-oxopropanoate |
| SMILES | CCOC(=O)CC(=O)[O-].[K+] |
Chemical Identifiers
| 6148-64-7 | |
| 170.205 | |
| WVUCPRGADMCTBN-UHFFFAOYSA-M | |
| 3446434 | |
| CCOC(=O)CC(=O)[O-].[K+] |
| C5H7KO4 | |
| MFCD00035603 | |
| ethyl potassium malonate, potassium 3-ethoxy-3-oxopropanoate, potassium ethyl malonate, potassium monoethyl malonate, monoethyl malonate potassium salt, malonic acid monoethyl ester potassium salt, monoethyl potassium malonate, ethyl malonate potassium salt, propanedioic acid, monoethyl ester, potassium salt, ethylpotassiummalonate | |
| potassium;3-ethoxy-3-oxopropanoate |
Specifications
| 6148-64-7 | |
| White | |
| MFCD00035603 | |
| ethyl potassium malonate, potassium 3-ethoxy-3-oxopropanoate, potassium ethyl malonate, potassium monoethyl malonate, monoethyl malonate potassium salt, malonic acid monoethyl ester potassium salt, monoethyl potassium malonate, ethyl malonate potassium salt, propanedioic acid, monoethyl ester, potassium salt, ethylpotassiummalonate | |
| CCOC(=O)CC(=O)[O-].[K+] | |
| 170.205 | |
| 170.21 | |
| Crystalline Powder |
| 200°C | |
| C5H7KO4 | |
| 25 g | |
| WVUCPRGADMCTBN-UHFFFAOYSA-M | |
| potassium;3-ethoxy-3-oxopropanoate | |
| 3446434 | |
| ≥98.0% (T) | |
| Monoethyl Potassium Malonate |
Safety and Handling
EINECSNumber : (2)-3893
TSCA : No