missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monoethyl Pimelate 98.0+%, TCI America™
$119.36 - $329.20
Chemical Identifiers
| CAS | 33018-91-6 |
|---|---|
| Molecular Formula | C9H16O4 |
| Molecular Weight (g/mol) | 188.223 |
| MDL Number | MFCD00801202 |
| InChI Key | NQYXFXWKKYGBNL-UHFFFAOYSA-N |
| Synonym | ethyl hydrogen pimelate, pimelic acid monoethyl ester, heptanedioic acid monoethyl ester, ethyl hydrogen heptane-1,7-dioate, ethyl hydroden pimelate, heptandioic acid monoethyl ester, heptanedioic acid 1-ethyl ester, heptanedioic acid, 1-ethyl ester |
| PubChem CID | 118383 |
| IUPAC Name | 7-ethoxy-7-oxoheptanoic acid |
| SMILES | CCOC(=O)CCCCCC(=O)O |
Chemical Identifiers
| 33018-91-6 | |
| 188.223 | |
| NQYXFXWKKYGBNL-UHFFFAOYSA-N | |
| 118383 | |
| CCOC(=O)CCCCCC(=O)O |
| C9H16O4 | |
| MFCD00801202 | |
| ethyl hydrogen pimelate, pimelic acid monoethyl ester, heptanedioic acid monoethyl ester, ethyl hydrogen heptane-1,7-dioate, ethyl hydroden pimelate, heptandioic acid monoethyl ester, heptanedioic acid 1-ethyl ester, heptanedioic acid, 1-ethyl ester | |
| 7-ethoxy-7-oxoheptanoic acid |
Specifications
| 33018-91-6 | |
| 162°C | |
| MFCD00801202 | |
| ethyl hydrogen pimelate, pimelic acid monoethyl ester, heptanedioic acid monoethyl ester, ethyl hydrogen heptane-1,7-dioate, ethyl hydroden pimelate, heptandioic acid monoethyl ester, heptanedioic acid 1-ethyl ester, heptanedioic acid, 1-ethyl ester | |
| CCOC(=O)CCCCCC(=O)O | |
| 188.223 | |
| 188.22 | |
| Liquid |
| Yellow | |
| C9H16O4 | |
| 5 g | |
| NQYXFXWKKYGBNL-UHFFFAOYSA-N | |
| 7-ethoxy-7-oxoheptanoic acid | |
| 118383 | |
| ≥98.0% (GC) | |
| Monoethyl Pimelate |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator