missing translation for 'onlineSavingsMsg'
Learn More

Monoethyl Pimelate 98.0+%, TCI America™

Catalog No. P12445G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P12445G 5 g
P124425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P12445G Supplier TCI America Supplier No. P12445G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 33018-91-6
Molecular Formula C9H16O4
Molecular Weight (g/mol) 188.223
MDL Number MFCD00801202
InChI Key NQYXFXWKKYGBNL-UHFFFAOYSA-N
Synonym ethyl hydrogen pimelate, pimelic acid monoethyl ester, heptanedioic acid monoethyl ester, ethyl hydrogen heptane-1,7-dioate, ethyl hydroden pimelate, heptandioic acid monoethyl ester, heptanedioic acid 1-ethyl ester, heptanedioic acid, 1-ethyl ester
PubChem CID 118383
IUPAC Name 7-ethoxy-7-oxoheptanoic acid
SMILES CCOC(=O)CCCCCC(=O)O

Specifications

Color Yellow
Boiling Point 162°C
Quantity 5 g
Formula Weight 188.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Monoethyl Pimelate
TSCA No
Recommended Storage Refrigerator