missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monoethyl Malonate, TCI America™
$67.38 - $67.38
Chemical Identifiers
| CAS | 1071-46-1 |
|---|---|
| Molecular Formula | C5H8O4 |
| Molecular Weight (g/mol) | 132.115 |
| MDL Number | MFCD00020490 |
| InChI Key | HGINADPHJQTSKN-UHFFFAOYSA-N |
| Synonym | ethyl hydrogen malonate, monoethyl malonate, monoethyl malonic acid, mono-ethyl malonate, monoethyl hydrogen malonate, malonic acid monoethyl ester, ethoxycarbonyl acetic acid, 2-ethoxycarbonyl acetic acid, 3-ethoxy-3-oxo-propanoic acid, ethylhydrogenmalonate |
| PubChem CID | 70615 |
| ChEBI | CHEBI:86907 |
| IUPAC Name | 3-ethoxy-3-oxopropanoic acid |
| SMILES | CCOC(=O)CC(=O)O |
Chemical Identifiers
| 1071-46-1 | |
| 132.115 | |
| HGINADPHJQTSKN-UHFFFAOYSA-N | |
| 70615 | |
| 3-ethoxy-3-oxopropanoic acid |
| C5H8O4 | |
| MFCD00020490 | |
| ethyl hydrogen malonate, monoethyl malonate, monoethyl malonic acid, mono-ethyl malonate, monoethyl hydrogen malonate, malonic acid monoethyl ester, ethoxycarbonyl acetic acid, 2-ethoxycarbonyl acetic acid, 3-ethoxy-3-oxo-propanoic acid, ethylhydrogenmalonate | |
| CHEBI:86907 | |
| CCOC(=O)CC(=O)O |
Specifications
| 1071-46-1 | |
| Yellow | |
| C5H8O4 | |
| 5 g | |
| HGINADPHJQTSKN-UHFFFAOYSA-N | |
| 3-ethoxy-3-oxopropanoic acid | |
| 70615 | |
| 132.12 | |
| Liquid |
| -13°C | |
| 208°C | |
| MFCD00020490 | |
| ethyl hydrogen malonate, monoethyl malonate, monoethyl malonic acid, mono-ethyl malonate, monoethyl hydrogen malonate, malonic acid monoethyl ester, ethoxycarbonyl acetic acid, 2-ethoxycarbonyl acetic acid, 3-ethoxy-3-oxo-propanoic acid, ethylhydrogenmalonate | |
| CCOC(=O)CC(=O)O | |
| 132.115 | |
| CHEBI:86907 | |
| ≥98.0% (T) | |
| Monoethyl Malonate |
Safety and Handling
EINECSNumber : (2)-0913&(2)-3691
TSCA : No