missing translation for 'onlineSavingsMsg'
Learn More

Monoethyl Malonate, TCI America™

Catalog No. M27005G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M27005G Supplier TCI America Supplier No. M27005G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1071-46-1
Molecular Formula C5H8O4
Molecular Weight (g/mol) 132.115
MDL Number MFCD00020490
InChI Key HGINADPHJQTSKN-UHFFFAOYSA-N
Synonym ethyl hydrogen malonate, monoethyl malonate, monoethyl malonic acid, mono-ethyl malonate, monoethyl hydrogen malonate, malonic acid monoethyl ester, ethoxycarbonyl acetic acid, 2-ethoxycarbonyl acetic acid, 3-ethoxy-3-oxo-propanoic acid, ethylhydrogenmalonate
PubChem CID 70615
ChEBI CHEBI:86907
IUPAC Name 3-ethoxy-3-oxopropanoic acid
SMILES CCOC(=O)CC(=O)O

Specifications

Melting Point -13°C
Color Yellow
Boiling Point 208°C
Quantity 5 g
Formula Weight 132.12
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Monoethyl Malonate
EINECSNumber (2)-0913&(2)-3691
TSCA No