missing translation for 'onlineSavingsMsg'
Learn More

Monoethyl Itaconate 97.0+%, TCI America™

Catalog No. p-7131301
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I026825G 25 g
I0268500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I026825G Supplier TCI America Supplier No. I026825G

Chemical Identifiers

CAS 57718-07-7
Molecular Formula C7H10O4
Molecular Weight (g/mol) 158.15
MDL Number MFCD00059138
InChI Key RTTAGBVNSDJDTE-UHFFFAOYSA-N
Synonym beta-Ethyl Itaconate, 4-Ethyl Methylenesuccinate, Itaconic Acid Monoethyl Ester
PubChem CID 533740
IUPAC Name 4-ethoxy-2-methylidene-4-oxobutanoic acid
SMILES CCOC(=O)CC(=C)C(O)=O

Specifications

Melting Point 56°C
Color White-Yellow
Boiling Point 153°C
Quantity 25 g
Formula Weight 158.15
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Monoethyl Itaconate
TSCA No
Recommended Storage Refrigerator