missing translation for 'onlineSavingsMsg'
Learn More

Monoethanolamine, NF, 98-100.5%, Spectrum™ Chemical

Catalog No. p-7157879
Change view
Click to view available options
Quantity:
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604680 4 L Amber Glass Bottle
18604681 20 L Poly Drum
18604682 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18604680 Supplier Spectrum Chemical Mfg Cor Supplier No. M13224LT
Only null left

M1322, 141-43-5, C2H7NO

Spectrum™ Chemical Monoethanolamine, NF - All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 141-43-5
Molecular Formula C2H7NO
Molecular Weight (g/mol) 61.08
MDL Number MFCD00008183
InChI Key HZAXFHJVJLSVMW-UHFFFAOYSA-N
IUPAC Name 2-aminoethan-1-ol
SMILES NCCO

Specifications

CAS Max % 1
Ignition Residue 0.001
Packaging Amber Glass Bottle
Quantity 4 L
Percent Purity 98 to 100.5%
Grade NF