Antibody;250TESTS;IgG1/kappa;Specifically recognizes the antigen-recognition domain of FMC63 derived CARs.;Monoclonal Anti-FMC63 scFv Antibody, Mouse IgG1 (Y45) (Cat. No. FM3-Y45P1) is expressed from human 293 cells.;This protein . ;2e5 of Anti-CD19 CAR-293 cells were stained with 100 L of 1:50 dilution (2 L stock solution in 100 L FACS buffer) Monoclonal Anti-FMC63 scFv Antibody, Mouse IgG1 (Cat. No. FM3-Y45P1) and isotype control respectively, washed and then followed by PE anti-mouse IgG1 antibody and analyzed with FACS (QC tested).