missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monochloroacetic Acid Buffer For Carbamate Analysis, Reagents
Specifications
Specifications
| Chemical Name or Material | Buffer |
| CAS | 7732-18-5 |
| Color | Colorless |
| pH | 3 |
| Physical Form | Liquid |
| Quantity | 1 L |
| Packaging | Glass Bottle |
| Sterility | Non-sterile |
| Solubility Information | Completely miscible |