missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monoammonium Glycyrrhizinate Hydrate 75.0+%, TCI America™
Supplier: TCI America G015125G
Specifications
| Monoammonium Glycyrrhizinate Hydrate | |
| 217°C | |
| C42H65NO16 | |
| 25 g | |
| ILRKKHJEINIICQ-OOFFSTKBSA-N | |
| (2S,3S,4S,5R,6S)-6-{[(3S,4aR,6aR,6bS,8aS,11S,12aR,14aR,14bS)-11-carboxy-4,4,6a,6b,8a,11,14b-heptamethyl-14-oxo-1,2,3,4,4a,5,6,6a,6b,7,8,8a,9,10,11,12,12a,14,14a,14b-icosahydropicen-3-yl]oxy}-5-{[(2R,3R,4S,5S,6S)-6-carboxy-3,4,5-trihydroxyoxan-2-yl]oxy}-3,4-dihydroxyoxane-2-carboxylic acid amine | |
| 131676200 | |
| ≥75.0% (HPLC) |
| 53956-04-0 | |
| White-Yellow | |
| MFCD00167400,MFCD00167400 | |
| Glycyrrhizic Acid Monoammonium Salt, Glycyrrhizin Monoammonium Salt | |
| N.CC1(C)[C@H](CC[C@@]2(C)[C@H]1CC[C@]1(C)[C@@H]2C(=O)C=C2[C@@H]3C[C@](C)(CC[C@]3(C)CC[C@@]12C)C(O)=O)O[C@H]1O[C@@H]([C@@H](O)[C@H](O)[C@H]1O[C@@H]1O[C@@H]([C@@H](O)[C@H](O)[C@H]1O)C(O)=O)C(O)=O | |
| 839.97 | |
| 839.97 | |
| Crystalline Powder |
Chemical Identifiers
| 53956-04-0 | |
| 839.97 | |
| ILRKKHJEINIICQ-OOFFSTKBSA-N | |
| 131676200 | |
| N.CC1(C)[C@H](CC[C@@]2(C)[C@H]1CC[C@]1(C)[C@@H]2C(=O)C=C2[C@@H]3C[C@](C)(CC[C@]3(C)CC[C@@]12C)C(O)=O)O[C@H]1O[C@@H]([C@@H](O)[C@H](O)[C@H]1O[C@@H]1O[C@@H]([C@@H](O)[C@H](O)[C@H]1O)C(O)=O)C(O)=O |
| C42H65NO16 | |
| MFCD00167400,MFCD00167400 | |
| Glycyrrhizic Acid Monoammonium Salt, Glycyrrhizin Monoammonium Salt | |
| (2S,3S,4S,5R,6S)-6-{[(3S,4aR,6aR,6bS,8aS,11S,12aR,14aR,14bS)-11-carboxy-4,4,6a,6b,8a,11,14b-heptamethyl-14-oxo-1,2,3,4,4a,5,6,6a,6b,7,8,8a,9,10,11,12,12a,14,14a,14b-icosahydropicen-3-yl]oxy}-5-{[(2R,3R,4S,5S,6S)-6-carboxy-3,4,5-trihydroxyoxan-2-yl]oxy}-3,4-dihydroxyoxane-2-carboxylic acid amine |
Safety and Handling
EINECSNumber : (9)-0398
RTECSNumber : LZ6500000
TSCA : Yes