missing translation for 'onlineSavingsMsg'
Learn More

Mono-tert-butyl Succinate 98.0+%, TCI America™

Catalog No. p-7133545
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M30331G 1 g
M30335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M30331G Supplier TCI America Supplier No. M30331G

Chemical Identifiers

CAS 15026-17-2
Molecular Formula C8H14O4
Molecular Weight (g/mol) 174.20
MDL Number MFCD00273441
InChI Key PCOCFIOYWNCGBM-UHFFFAOYSA-N
Synonym mono-tert-butyl succinate, 4-tert-butoxy-4-oxobutanoic acid, succinic acid mono-tert-butyl ester, tert-butyl hydrogen succinate, mono-t-butyl succinate, pubchem11616, succinic acid 1-tert-butyl ester, succinic acid 4-tert-butyl ester
PubChem CID 4139107
IUPAC Name 4-(tert-butoxy)-4-oxobutanoic acid
SMILES CC(C)(C)OC(=O)CCC(O)=O

Specifications

Melting Point 53°C
Color White
Boiling Point 92°C
Quantity 1 g
Formula Weight 174.20
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Mono-tert-butyl Succinate