missing translation for 'onlineSavingsMsg'
Learn More

Mono-tert-butyl Malonate 97.0+%, TCI America™

Catalog No. M26875G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M26875G 5 g
M268725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M26875G Supplier TCI America Supplier No. M26875G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 40052-13-9
Molecular Formula C7H12O4
Molecular Weight (g/mol) 160.169
MDL Number MFCD00191886
InChI Key NGGGZUAEOKRHMA-UHFFFAOYSA-N
Synonym 3-tert-butoxy-3-oxopropanoic acid, malonic acid mono-tert-butyl ester, mono-tert-butyl malonate, 2-tert-butoxycarbonyl acetic acid, tert-butyl hydrogen malonate, mono-t-butyl malonate, pubchem13754, malonicacidmono-tert-butylester, malonic acid mono t-butyl ester
PubChem CID 545853
IUPAC Name 3-[(2-methylpropan-2-yl)oxy]-3-oxopropanoic acid
SMILES CC(C)(C)OC(=O)CC(=O)O

Specifications

Melting Point 19°C
Color Yellow
Boiling Point 90°C
Quantity 5 g
Formula Weight 160.17
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Mono-tert-butyl Malonate