missing translation for 'onlineSavingsMsg'
Learn More

Mono-Poly™ Resolving Medium

Catalog No. ICN1698049
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN1698049 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN1698049 Supplier MP Biomedicals Inc Supplier No. 091698049

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mono-Poly™ Resolving Medium separates mononuclear and polymorphonuclear leukocytes from whole human blood in two distinct bands.

Mono-Poly is highly versatile and can be used for a variety of research and clinical applications such as histocompatibility testing, lymphocyte studies, in vitro cell mediated immunity assays, or any procedure requiring pure white blood cell preparation

Product provides:

  • Separation of mononuclear and polymorphonuclear leukocytes in two distinct bands
  • Greater than 95% cell viability
  • Easy, single step procedure
  • No sample dilution requirement or lysis of red blood cells

Specifications

Quantity 100 mL
Product Type Mono-Poly™ Resolving Medium