missing translation for 'onlineSavingsMsg'
Learn More

mono-n-Dodecyl phosphate, tech. 90%

Catalog No. AAL1194114
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1194114 25 g
AAL1194106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1194114 Supplier Thermo Scientific Chemicals Supplier No. L1194114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2627-35-2 | C12H27O4P | 266.32 g/mol

mono-n-Dodecyl phosphate is a lysophosphatidic acid (LPA) receptor ligand which serves as a specific agonist at EDG-4 (LPA2) (EC50=700 nM) and antagonist at EDG-7

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
mono-n-Dodecyl phosphate is a lysophosphatidic acid (LPA) receptor ligand which serves as a specific agonist at EDG-4 (LPA2) (EC50=700 nM) and antagonist at EDG-7

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases.

Chemical Identifiers

CAS 2627-35-2
Molecular Formula C12H27O4P
Molecular Weight (g/mol) 266.32
MDL Number MFCD00013946
InChI Key TVACALAUIQMRDF-UHFFFAOYSA-N
Synonym dodecyl phosphate, monolauryl phosphate, phosphoric acid, monododecyl ester, mono-n-dodecyl phosphate, monododecyl phosphate, dihydrogen monolauryl phosphate, lauryl dihydrogen phosphate, unii-0k08l8ge2h, phosphoric acid monododecyl ester, dodecyloxyphosphonic acid
PubChem CID 75816
IUPAC Name dodecyl dihydrogen phosphate
SMILES CCCCCCCCCCCCOP(O)(O)=O

Specifications

Melting Point 39°C to 45°C
Odor Odorless
Quantity 25 g
Beilstein 1710518
Solubility Information Insoluble in water.
Formula Weight 266.32
Percent Purity 90%
Grade Technical
Chemical Name or Material mono-n-Dodecyl phosphate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 220-095-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only