missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mono-Ethyl malonate, 95%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| CAS Min % | 94.0 |
| CAS Max % | 100.0 |
| Density | 1.1190g/mL |
| Boiling Point | 106.5°C (3.0 mmHg) |
| Assay Percent Range | 95% |
| Packaging | Glass bottle |
| Linear Formula | C2H5O2CCH2CO2H |
| Quantity | 25 g |
| Beilstein | 02,IV,188 |
| Specific Gravity | 1.119 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Causes skin irritation. May cause respiratory irritation. Causes serious eye irritation. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. If skin irritation occurs: Get medical advice/attention. If eye irritation persists: Get medical advice/attention. Avoid breathing dust/fume/ |
| EINECSNumber | 213-992-7 |
RUO – Research Use Only