missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mono-6-O-(p-toluenesulfonyl)-gamma-cyclodextrin 90.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 200 mg |
| Formula Weight | 1451.31 |
| Percent Purity | ≥90.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Mono-6-O-(p-toluenesulfonyl)-gamma-cyclodextrin |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |