missing translation for 'onlineSavingsMsg'
Learn More

Mono-4-nitrobenzyl Malonate 98.0+%, TCI America™

Catalog No. M11775G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M11775G 5 g
M117725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M11775G Supplier TCI America Supplier No. M11775G
Only null left

Chemical Identifiers

CAS 77359-11-6
Molecular Formula C10H9NO6
Molecular Weight (g/mol) 239.183
MDL Number MFCD00191556
InChI Key RIGFMUNSTCPGNP-UHFFFAOYSA-N
Synonym Malonic Acid Mono-4-nitrobenzyl Ester, 4-Nitrobenzyl Hydrogen Malonate
PubChem CID 592642
SMILES C1=CC(=CC=C1COC(=O)CC(=O)O)[N+](=O)[O-]

Specifications

Melting Point 109°C
Color White-Yellow
Quantity 5 g
Formula Weight 239.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Mono-4-nitrobenzyl Malonate
EINECSNumber (3)-4274
TSCA No