missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mono-2-O-(p-toluenesulfonyl)-gamma-cyclodextrin 95.0+%, TCI America™
Specifications
Specifications
| CAS | 97227-32-2 |
| Color | White-Yellow |
| Molecular Formula | C55H86O42S |
| Quantity | 1 g |
| InChI Key | USRILNNILPQZNC-SCLKBCEUSA-N |
| SMILES | CC1=CC=C(C=C1)S(=O)(=O)OC2C(C3C(OC2OC4C(OC(C(C4O)O)OC5C(OC(C(C5O)O)OC6C(OC(C(C6O)O)OC7C(OC(C(C7O)O)OC8C(OC(C(C8O)O)OC9C(OC(C(C9O)O)OC1C(OC(O3)C(C1O)O)CO)CO)CO)CO)CO)CO)CO)CO)O |
| Molecular Weight (g/mol) | 1451.311 |
| PubChem CID | 11018749 |
| Formula Weight | 1451.31 |
| Percent Purity | ≥95.0% (HPLC) |
| Show More |
Safety and Handling
| TSCA | No |