missing translation for 'onlineSavingsMsg'
Learn More

Mono-2-O-(p-toluenesulfonyl)-beta-cyclodextrin Hydrate 97.0+%, TCI America™

Catalog No. M1741200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1741200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1741200MG Supplier TCI America Supplier No. M1741200MG
Only null left

Specifications

CAS 84216-71-7
Color White
Molecular Formula C49H76O37S
Quantity 200 mg
InChI Key VGZWDYWXWGKKEE-UJPGXMRNSA-N
SMILES CC1=CC=C(C=C1)S(=O)(=O)OC2C(C3C(OC2OC4C(OC(C(C4O)O)OC5C(OC(C(C5O)O)OC6C(OC(C(C6O)O)OC7C(OC(C(C7O)O)OC8C(OC(C(C8O)O)OC9C(OC(O3)C(C9O)O)CO)CO)CO)CO)CO)CO)CO)O
Molecular Weight (g/mol) 1289.17
PubChem CID 11829278
Formula Weight 1289.17
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Mono-2-O-(p-toluenesulfonyl)-beta-cyclodextrin Hydrate
Show More Show Less