missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mono(2-acryloyloxyethyl) Succinate (stabilized with MEHQ) 90.0+%, TCI America™
$54.06 - $270.29
Chemical Identifiers
| CAS | 50940-49-3 |
|---|---|
| Molecular Formula | C9H12O6 |
| Molecular Weight (g/mol) | 216.189 |
| MDL Number | MFCD00274301 |
| InChI Key | UZDMJPAQQFSMMV-UHFFFAOYSA-N |
| Synonym | Succinic Acid Mono(2-acryloyloxyethyl) Ester, 2-(Acryloyloxy)ethyl Hydrogen Succinate |
| PubChem CID | 175956 |
| IUPAC Name | 4-oxo-4-(2-prop-2-enoyloxyethoxy)butanoic acid |
| SMILES | C=CC(=O)OCCOC(=O)CCC(=O)O |
Chemical Identifiers
| 50940-49-3 | |
| 216.189 | |
| UZDMJPAQQFSMMV-UHFFFAOYSA-N | |
| 175956 | |
| C=CC(=O)OCCOC(=O)CCC(=O)O |
| C9H12O6 | |
| MFCD00274301 | |
| Succinic Acid Mono(2-acryloyloxyethyl) Ester, 2-(Acryloyloxy)ethyl Hydrogen Succinate | |
| 4-oxo-4-(2-prop-2-enoyloxyethoxy)butanoic acid |
Specifications
| 50940-49-3 | |
| MFCD00274301 | |
| Succinic Acid Mono(2-acryloyloxyethyl) Ester, 2-(Acryloyloxy)ethyl Hydrogen Succinate | |
| C=CC(=O)OCCOC(=O)CCC(=O)O | |
| 216.189 | |
| 216.19 | |
| Liquid |
| C9H12O6 | |
| 25 g | |
| UZDMJPAQQFSMMV-UHFFFAOYSA-N | |
| 4-oxo-4-(2-prop-2-enoyloxyethoxy)butanoic acid | |
| 175956 | |
| ≥90.0% (GC) | |
| Mono(2-acryloyloxyethyl) Succinate (stabilized with MEHQ) |
Safety and Handling
EINECSNumber : (2)-1006
TSCA : Yes