missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mono(2-acryloyloxyethyl) Succinate (stabilized with MEHQ) 90.0+%, TCI America™
Specifications
Specifications
| Quantity | 25 g |
| Formula Weight | 216.19 |
| Percent Purity | ≥90.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Mono(2-acryloyloxyethyl) Succinate (stabilized with MEHQ) |
Safety and Handling
| EINECSNumber | (2)-1006 |
| TSCA | Yes |