missing translation for 'onlineSavingsMsg'
Learn More

Bio-Serv™ Monkey Shine Mirrors

Catalog No. 14727058
Encompass
Change view
Click to view available options
For Use With (Equipment):
With chain assembly
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
14727058 With chain assembly
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14727058 Supplier Bio-Serv™ Supplier No. K3150

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

  • Made of durable stainless steel
  • Shiny on both sides
  • Smoothed edges for safety
  • Available without chain assembly for use inside the cage
  • Autoclavable
  • Cagewasher safe

Specifications

Product Type Primate Toy
Color Shiny Silver
Material Stainless Steel
For Use With (Equipment) With chain assembly
Diameter (English) Inside 6 in.