missing translation for 'onlineSavingsMsg'
Learn More

Monkey (cynomolgus) S9 Fractions

Catalog No. MKS9PL
Encompass
Catalog No. MKS9PL Supplier Thermo Fisher Scientific Supplier No. MKS9PL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Concentration 20 mg⁄ml
Content And Storage Store between -60°C and -85°C.
Form Frozen
Gender Male
Species Monkey
Tissue Liver
Donor Source Pooled
Format Tube
Product Line Gibco
Product Type Monkey S9 Fraction
Quantity 1 mL
Shipping Condition Dry Ice
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.