missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monel∣r 400 gauze, 200 mesh woven from 0.05mm (0.0021in) dia wire
$136.29 - $136.29
Specifications
| Chemical Name or Material | Monel™ 400 gauze |
|---|---|
| Name Note | 200 mesh woven from 0.05mm (0.0021 in.) dia. wire |
| Mesh Size | 200 Mesh |
| MDL Number | MFCD09910536 |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Monel™ 400 gauze | |
| 200 mesh woven from 0.05mm (0.0021 in.) dia. wire | |
| 200 Mesh | |
| MFCD09910536 |
RUO – Research Use Only